Anti-DKK2 Goat Polyclonal Antibody

Supplier: Prosci
45-484
10115-908EA 701.94 USD
10115-908
Anti-DKK2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1: 8,000. Western Blot: Approximately 28 kDa band observed in human heart lysate (calculated MW of 28.5 kDa according to NP_055236). Recommended concentration: 1-3 ?g/mL

DKK2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DKK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR