Anti-DGAT1 Goat Polyclonal Antibody

Supplier: Prosci
45-475
10115-890EA 701.94 USD
10115-890
Anti-DGAT1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 50 kDa band observed in human duodenum lysate (calculated MW of 55.3 kDa according to NP_036211.1). Recommended concentration: 0.5-1 ?g/mL.

DGAT1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DGAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR