Anti-DEF6 Goat Polyclonal Antibody

Supplier: Prosci
45-473
10115-888EA 701.94 USD
10115-888
Anti-DEF6 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Approximately 70 kDa band observed in human thymus lysates (calculated MW of 73.9 kDa according to NP_071330.3). Recommended concentration: 1-3 ?g/mL.

DEF6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DEF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR