Anti-DDAH1 Goat Polyclonal Antibody

Supplier: Prosci
45-468
10115-880EA 701.94 USD
10115-880
Anti-DDAH1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 37 kDa band observed in human kidney, liver and pancreas lysates (calculated MW of 31.1 kDa according to NP_036269.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Pullamsetti et al, FASEB J. 2005 Jul;19 (9) :1175-7; PMID: 15827267). Recommended concentration: 0.1-0.3 ?g/mL.

DDAH1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DDAH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR