Anti-DCDC2 Goat Polyclonal Antibody

Supplier: Prosci
45-464
10115-872EA 701.94 USD
10115-872
Anti-DCDC2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: full length recombinant human DCDC2 expressed in COS7 (Lane A), non-transfectant COS7 (lane B). Approximately 52 kDa band observed (calculated MW of 52.8 kDa according to NP_057440.1). Recommmended concentration 0.1-0.3 ?g/mL.

DCDC2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DCDC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR