Anti-DAZL Goat Polyclonal Antibody

Supplier: Prosci
45-462
10115-868EA 701.76 USD
10115-868
Anti-DAZL Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 35 kDa band observed in human ovary and human testes lysates (calculated MW of 33.2 kDa according to NP_001342.2). Recommended concentration: 0.1-0.3 ?g/mL. Immunohistochemistry: In paraffin embedded mouse testis shows cytoplasm staining of spermatogonia. Additional staining is observed in the cytoplasm of elongatng spermatids but this additional staining is not in agreement with results obtained by another antibody. Recommended concentration, 1-3 ?g/mL.

DAZL antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DAZL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR