Anti-CYP17A1 Goat Polyclonal Antibody

Supplier: ProSci
45-446
10115-840EA 701.76 USD
10115-840
Anti-CYP17A1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 60 kDa band observed in human ovary lysates (calculated MW of 57.4 kDa according to NP_000093.1). Recommended concentration: 0.1-0.3 ?g/mL.

CYP17A1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CYP17A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR