Anti-CXCR3 Goat Polyclonal Antibody

Supplier: ProSci
45-439
10115-830EA 701.76 USD
10115-830
Anti-CXCR3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 50-55 kDa band in lysates of human thymus, kidney and placenta, and of cell line HeLa after 0.3 ?g/mL antibody staining. The 50-55 kDa band was successfully blocked by incubation with the immunizing peptide.

CXCR3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CXCR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR