Anti-CSNK1E Goat Polyclonal Antibody

Supplier: ProSci
45-437
10115-826EA 701.94 USD
10115-826
Anti-CSNK1E Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 45 kDa band observed in HEK293 lysate (calculated MW of 47.3 kDa according to NP_001885.1 and NP_689407.1). In transfected HEK293 transiently expressing CSNK1E a band of approximately 49 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.3-1 ?g/mL.

CSNK1E antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CSNK1E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR