Anti-NLRP3 Goat Polyclonal Antibody

Supplier: ProSci
45-435
10113-270EA 701.94 USD
10113-270
Anti-NLRP3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution >1:32000. Western Blot: No signal obtained yet but low background observed in Daudi, A431, Jurkat, U937 and MOLT-4 lysates at up to 1 ?g/mL.

Cryopyrin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NLRP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR