Anti-CRHR1 Goat Polyclonal Antibody

Supplier: ProSci
45-431
10115-818EA 701.94 USD
10115-818
Anti-CRHR1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Preliminary experiments in human colon, duodenum and placenta lysates gave no specific signal but low background (at antibody concentration up to 1 ?g/mL). Immunohistochemistry: this antibody is successfully used in immunohistochemistry on mouse brain. Immunofluorescence: this antibody is successfully used in immunofluoresence on mouse brain.

CRHR1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CRHR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR