Anti-CREB3L4 Goat Polyclonal Antibody

Supplier: ProSci
45-428
10115-814EA 701.94 USD
10115-814
Anti-CREB3L4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution >1:32,000. Western Blot: Approximately 50 kDa band observed in human placenta lysate (predicted MW of 45 kDa according to NP_570968). Recommended for use at 0.5-2 ?g/mL. Immunohistochemistry: In paraffin embedded human prostate shows strong staining of cytoplasm in the secretory cells of the gland. Recommended concentration, 2-4 ?g/mL.

CREB3L4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CREB3L4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR