Anti-CORO1C Goat Polyclonal Antibody

Supplier: ProSci
45-422
10115-806EA 701.94 USD
10115-806
Anti-CORO1C Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 55 kDa band observed in human lung and placenta lysates (calculated MW of 53.2 kDa according to NP_055140.1). Recommended for use at 0.2-1 ?g/mL.

Coronin 3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CORO1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR