Anti-COPS2 Goat Polyclonal Antibody

Supplier: ProSci

45-418
10115-800EA 560.36 USD
10115-800
Anti-COPS2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 48 kDa band observed in human brain, mouse brain lysates and in lysates of mouse putuitary cell line NIH3T3 (calculated MW of 51.6 kDa according to NP_004227.1). Recommended concentration: 1-3 ?g/mL. Immunohistochemistry: In paraffin embedded human thyroid gland shows nclear and cytoplasm staining in activated epithelial cells Recommended concentration, 3-5 ?g/mL.

COPS2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: COPS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR