Anti-CARD16 Goat Polyclonal Antibody

Supplier: ProSci
45-417
10115-798EA 701.94 USD
10115-798
Anti-CARD16 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 28-30 kDa band in human colon, pancreas, placenta and tonsil lysates after 0.3 ?g/mL antibody staining.7 kDa according to NP_443121.1. The 28-30 kDa band was successfully blocked by incubation with the immunizing peptide.

COP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CARD16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR