Anti-DPP4 Goat Polyclonal Antibody

Supplier: ProSci
45-381
10115-744EA 701.94 USD
10115-744
Anti-DPP4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Approximately 110 kDa band observed in rat lung lysates (calculated MW of 88.3Da according to NP_001926.2). The observed molecular weight of this glycoprotein corresponds to earlier findings in literature with different antibodies (Herrera et al, J Biol Chem. 2001 Jun 1;276 (22) :19532-9.; PMID: 11278278). Recommended concentration: 0.3-1 ?g/mL.

CD26 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DPP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR