Anti-CCL3L1 Goat Polyclonal Antibody

Supplier: ProSci
45-379
10115-740EA 552.17 USD
10115-740
Anti-CCL3L1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 26+28 kDa (doublet) and 40 kDa in human bone marrow and spleen lysates after 0.1 ?g/mL antibody staining.The 26+28 kDa doublet was also found in human lymph node and in tonsil lysates.2 kDa according to NP_066286.1. The detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain these bands).

CCL3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CCL3L1; CCL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR