Anti-CCKAR Goat Polyclonal Antibody

Supplier: ProSci
45-377
10115-736EA 701.94 USD
10115-736
Anti-CCKAR Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4000. Western Blot: Preliminary experiments in human pancreas lysates gave no specific signal but low background (at antibody concentration up to 1 ?g/mL). Immunohistochemistry: In paraffin embedded human pancreas, shows cytoplasmic staining of some cells in the islets of Langerhans. Recommended concentration, 10 ?g/mL.

CCKAR antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CCKAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR