Anti-CBX6 / CBX8 Goat Polyclonal Antibody

45-373
10115-728EA 389.35 USD
10115-728
Anti-CBX6 / CBX8 Goat Polyclonal Antibody
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 37 kDa band in human, mouse and rat brain lysates at 0.3 ?g/mL.8 kDa according to NP_055107.

CBX6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CBX6 / CBX8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR