Anti-CBX1 Goat Polyclonal Antibody

Supplier: ProSci
45-370
10115-722EA 701.94 USD
10115-722
Anti-CBX1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 25 kDa band observed in lysates of A431, HeLa and HepG2 (calculated MW of 25 kDa according to NP_006798.1). Recommended concentration: 0.3-1.0 ?g/mL. Immunohistochemistry: In paraffin embedded human placenta shows strong nuclear staining in trophoblasts. Recommended concentration, 2-3 ?g/mL.

CBX1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CBX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR