Anti-CBR1 Goat Polyclonal Antibody

Supplier: ProSci
45-368
10115-718EA 701.94 USD
10115-718
Anti-CBR1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 30-35 kDa band seen in human liver and kidney lysates [Calculated MW of 30.4 kDa according to NP_001748.1]. Recommended for use at 0.2-0.5 ?g/mL. Immunohistochemistry: In paraffin embedded human kidney shows cytoplasmic staining of epithelial cells of renal tubules. Recommended concentration, 1-2 ?g/mL.

CBR1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CBR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR