Anti-CTNNA1 Goat Polyclonal Antibody

Supplier: ProSci

45-364
10115-712EA 560.36 USD
10115-712
Anti-CTNNA1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 100 kDa band observed in human colon and human skin lysates (calculated MW of 100 kDa according to NP_001894.2). Recommended concentration: 1-3 ?g/mL. Immunohistochemistry: In paraffin embedded human prostate shows staining of cell membranes of the secretory cells in the glands. Recommended concentration, 3-6 ?g/mL.

Catenin alpH a 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CTNNA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR