Anti-CARD6 Goat Polyclonal Antibody

Supplier: ProSci
45-361
10115-706EA 701.94 USD
10115-706
Anti-CARD6 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution >1:32,000. Western Blot: Approximately 45-50 kDa and 25-30 kDa bands observed in A549, human heart and human muscle lysates at 1 ?g of primary. Please note that the bands we observe are different from the predicted size of approximately 119 kDa according to NP_115976. Immunohistochemistry: In paraffin embedded human pancreas shows variable staining of cytoplasm in acinar cells. Recommended concentration, 5-10 ?g/mL.

CARD6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CARD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR