Anti-C14ORF68 Goat Polyclonal Antibody

Supplier: ProSci
45-347
10113-250EA 701.76 USD
10113-250
Anti-C14ORF68 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 22 kDa band in human liver lysates after 0.5 ?g/mL antibody staining.4 kDa according to NP_997000.2. The 22 kDa band was successfully blocked by incubation with the immunizing peptide.

C14orf68 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: C14orf68
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR