Anti-CKB Goat Polyclonal Antibody

Supplier: ProSci
45-339
10115-678EA 701.94 USD
10115-678
Anti-CKB Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 45-48 kDa band observed in human brain lysates (calculated MW of 42.6 kDa according to NP_001814.2). Recommended concentration: 0.1-0.3 ?g/mL.

Brain Creatine Kinase antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR