Anti-BNIP1 Goat Polyclonal Antibody

Supplier: ProSci
45-335
10115-670EA 701.94 USD
10115-670
Anti-BNIP1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 28 kDa band observed in human kidney lysates (calculated MW of 26.1 kDa according to NP_001196.1; NP_053581.1; NP_053582.1; NP_053583.1). Recommended concentration: 0.5-1.5 ?g/mL. an additional band of unknown identity was also consistently observed at 48 kDa . This band was successfully blocked by incubation with the immunizing peptide.

BNIP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BNIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR