Anti-BIN1 Goat Polyclonal Antibody

Supplier: ProSci
45-332
10115-666EA 701.94 USD
10115-666
Anti-BIN1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: experiments gave bands at approximately 70 kDa and 60 kDa in human skeletal muscle lysates after 0.1 ?g/mL antibody staining. These bands correspond to earlier findings in literature with different antibodies (Chang et al, tumor Res. 2007 Jan 1;67 (1) :100-7.; PMID: 17210688). This protein has a calculated MW of 64.7 kDa according to NP_647593.1. Recommended concentration: 1-3 ?g/mL.

BIN1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR