Anti-BAZ2B Goat Polyclonal Antibody

Supplier: ProSci
45-322
10115-650EA 701.94 USD
10115-650
Anti-BAZ2B Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 35 kDa band in testis and brain lysates after 0.3 ?g/mL antibody staining. The 35 kDa band was successfully blocked by incubation with the immunizing peptide. Immunohistochemistry: In paraffin embedded human small intestine shows primarily nuclear staining in a number of cells of the lamina propria. Recommended concentration, 1-2 ?g/mL.

BAZ2B antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BAZ2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR