Anti-SMARCE1 Goat Polyclonal Antibody

Supplier: ProSci
45-315
10112-492EA 701.94 USD
10112-492
Anti-SMARCE1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 50 kDa band observed in lysates of cell lines HEPG2, HeLa and Jurkat (calculated MW of 46.6 kDa according to NP_003070.3). A 48 kDa band was observed in transfected HEK293 transiently expressing BAF57. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.2-0.6 ?g/mL.

BAF57 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SMARCE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR