Anti-PPP2R5B Goat Polyclonal Antibody

Supplier: ProSci
45-312
10112-486EA 701.94 USD
10112-486
Anti-PPP2R5B Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Approximately 55 kDa band observed in human cerebellum lysates (calculated MW of 57.4 kDa according to NP_006235.1). Recommended concentration: 2-3 ?g/mL.

B56 beta antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PPP2R5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR