Anti-TBC1D4 Goat Polyclonal Antibody

Supplier: ProSci
45-298
10112-470EA 701.94 USD
10112-470
Anti-TBC1D4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 150 kDa band observed in lysates of cell lines Daudi and MOLT4 (calculated MW of 147 kDa according to NP_055647.2). Recommended concentration: 0.1-0.3 ?g/mL. Immunohistochemistry: In paraffin embedded human skeletal muscle shows striated pattern staining of majority of muscular fibres. Recommended concentration, 3-5 ?g/mL.

AS160 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TBC1D4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR