Anti-ARPC3 Goat Polyclonal Antibody

Supplier: ProSci

45-294
10112-462EA 560.36 USD
10112-462
Anti-ARPC3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Band observed just below 20 kDa marker in Hela, human brain and mouse brain lysates (predicted MW of 20 kDa according to NP_005710). Recommended for use at 1-3 ?g/mL.

ARPC3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ARPC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR