Anti-ACTR1B Goat Polyclonal Antibody

Supplier: ProSci
45-291
10113-230EA 701.94 USD
10113-230
Anti-ACTR1B Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 45 kDa band observed in human heart and brain lysates (predicted MW of 43 kDa according to NP_005726). Recommended for use at 0.5-2 ?g/mL. Immunohistochemistry: In paraffin embedded human uterus shows fibrillar staining of cytoplasm in endometrial cells. Recommended concentration, 3-5 ?g/mL.

ARP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ACTR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR