Anti-ARL2 Goat Polyclonal Antibody

Supplier: ProSci
45-285
10112-450EA 701.94 USD
10112-450
Anti-ARL2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:1000. Western Blot: Approximately 20 kDa band seen in U937 lysate (Expected MW from amino acid sequence according to NP_001658 is 20 kDa ). Recommended for use at 1-3 ?g/mL.

ARL2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ARL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR