Anti-ARG1 Goat Polyclonal Antibody

Supplier: ProSci
45-280
10113-226EA 701.94 USD
10113-226
Anti-ARG1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 38 kDa band observed in mouse liver and rat liver lysates (calculated MW of 35.0 kDa according to NP_058830.1). Recommended concentration: 0.03-0.3 ?g/mL.

ARG1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Arg1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR