Anti-ARFGAP3 Goat Polyclonal Antibody

Supplier: ProSci
45-278
10112-442EA 701.94 USD
10112-442
Anti-ARFGAP3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 55-60 kDa band seen in mouse brain extracts. Recommended for use at 1-3 ?g/mL.

ARFGAP3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ARFGAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR