Anti-APOA4 Goat Polyclonal Antibody

Supplier: ProSci
45-263
10112-420EA 701.94 USD
10112-420
Anti-APOA4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 45 kDa band observed in human duodenum lysates (calculated MW of 45.4 kDa according to NP_000473.2). Recommended concentration: 0.01-0.03 ?g/mL.

APOA4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: APOA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR