Anti-APBB1 Goat Polyclonal Antibody

Supplier: ProSci
45-261
10112-416EA 701.94 USD
10112-416
Anti-APBB1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128,000. Approximately 100 kDa band observed in lysates of NIH/3T3 and HeLa (calculated MW of 77.2 kDa according to NP_001155.1 and 77.0 kDa according to NP_663722.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Zambrano et al, J Biol Chem. 1998 Aug 7;273 (32) :20128-33.; PMID: 9685356). Recommended concentration: 0.1-0.3 ?g/mL.

APBB1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: APBB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR