Anti-ANXA10 Goat Polyclonal Antibody

Supplier: ProSci
45-255
10112-406EA 701.76 USD
10112-406
Anti-ANXA10 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: At 1 ?g/mL antibody banded at approximately 45-48 kDa in Hela, A431 and human kidney lysates. Immunohistochemistry: In paraffin embedded human pancreas shows staining of islet of Langerhans. Recommended concentration, 3-5 ?g/mL.

Annexin A10 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ANXA10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR