Anti-NIF3L1 Goat Polyclonal Antibody

Supplier: ProSci
45-245
10112-392EA 701.94 USD
10112-392
Anti-NIF3L1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 40 kDa band seen in human heart and human testis extracts [Predicted MW of approximately 39 kDa according to NP_068596]. Recommended for use at 0.2-1 ?g/mL. Immunohistochemistry: In paraffin embedded human breast shows strong cytoplasm staining in epithelial and myoepithelial duct cells. Recommended concentration, 2-4 ?g/mL.

ALS2CR1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NIF3L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR