Anti-ALOX5AP Goat Polyclonal Antibody

Supplier: ProSci
45-241
10113-214EA 701.94 USD
10113-214
Anti-ALOX5AP Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 48 kDa band in human brain and U937 lysates at 1 ?g/mL. Further, this band was successfully blocked by incubation with the immunizing peptide. ALOX5AP antibody has been successfully used in WB assays with transfected HeLa cells (customer feedback, see Maier TJ et al., 2008).

ALOX5AP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ALOX5AP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR