Anti-AKAP9 Goat Polyclonal Antibody

Supplier: ProSci
45-232
10112-374EA 701.94 USD
10112-374
Anti-AKAP9 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 450 kDa band observed in HEPG2 lysate (calculated MW of 454 kDa according to NP_ 671700.1). Recommended concentration: 1-3 ?g/mL.

AKAP9 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AKAP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR