Anti-AKAP10 Goat Polyclonal Antibody

Supplier: ProSci
45-228
10112-366EA 701.94 USD
10112-366
Anti-AKAP10 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 74 kDa band observed in rat brain lysates (calculated MW of 73.8 kDa according to human NP_009133.2 and according to rat NP_001108078.1). Recommended concentration: 0.3-1 ?g/mL.

AKAP10 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AKAP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR