Anti-AIF1 Goat Polyclonal Antibody

Supplier: ProSci
45-221
10113-202EA 701.94 USD
10113-202
Anti-AIF1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 16 kDa band observed in human lymph node and spleen lysates (calculated MW of 16.7 kDa according to NP_001614.3. Recommended concentration: 1-3 ?g/mL. Immunohistochemistry: In paraffin embedded human placenta shows strong staining of extracellualr material (with both 45-220 and 45-221). Recommended concentration, 2-3 ?g/mL.

AIF1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR