Anti-AGTR1 Goat Polyclonal Antibody

Supplier: ProSci
45-216
10112-354EA 701.94 USD
10112-354
Anti-AGTR1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 38 kDa band observed in liver lysates of human, mouse and rat (calculated MW of 41.1 kDa according to NP_000676.1, NP_004826.2, NP_033611.1, NP_114038.1 and NP_114438.1). Recommended concentration: 0.3-1 ?g/mL.

AGTR1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AGTR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR