Anti-ADH7 Goat Polyclonal Antibody

Supplier: ProSci
45-208
10112-340EA 552.17 USD
10112-340
Anti-ADH7 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 26 kDa band in human duodenum lysate and liver and of rat stomach after 0.2 ?g/mL antibody staining.5 kDa according to NP_000664.2. The 26 kDa band was successfully blocked by incubation with the immunizing peptide.

ADH7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ADH7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR