Anti-ADORA2 Goat Polyclonal Antibody

Supplier: ProSci
45-205
10115-646EA 701.94 USD
10115-646
Anti-ADORA2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000.

Adenosine A2 alpH a Receptor antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ADORA2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rhesus monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR