Anti-ACTA2 Goat Polyclonal Antibody

Supplier: ProSci
45-198
10115-632EA 701.76 USD
10115-632
Anti-ACTA2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4000. Western Blot: Approximately 45 kDa band observed in human duodenum lysates (calculated MW of 42.0 kDa according to NP_001604.1). Recommended concentration: 0.1-0.3 ?g/mL.

Actin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ACTA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR