Anti-ACOX2 Goat Polyclonal Antibody

Supplier: ProSci
45-194
10113-194EA 701.94 USD
10113-194
Anti-ACOX2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 75 kDa band observed in human liver lysates (calculated MW of 76.8 kDa according to NP_003491.1). Recommended concentration: 0.5-1.5 ?g/mL. an additional band of 37 kDa was consistently observed, however this band was not blocked by the immunizing peptide and it is therefore a non-specific signal. We call for caution when used for other assays than Western Blot.

ACOX2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ACOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR