Anti-ACADM Goat Polyclonal Antibody

Supplier: ProSci
45-190
10115-622EA 701.94 USD
10115-622
Anti-ACADM Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 45 kDa band observed in human kidney and heart lysates (calculated MW of 46.6 kDa according to NP_000007.1). Recommended for use at 0.01-0.1 ?g/mL. Immunohistochemistry: In paraffin embedded human liver shows mitochondrial staining in hepatocytes. Recommended concentration, 3-5 ?g/mL.

ACADM antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ACADM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR